siRNA Details
Gene Symbol: Tmem156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025138.1 | NP_001020309.1 | 4 | 686 | ||
XM_008770160.1 | XP_008768382.1 | 5 | 901 | |||
GenBank mRNA | BC091217.1 | - | 4 | 686 | 7384360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
