siRNA Details
Gene Symbol: Hormad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017501.1 | NP_001017501.1 | 3 | 378 | ||
XM_008770340.1 | XP_008768562.1 | 3 | 473 | |||
XM_008770341.1 | XP_008768563.1 | 5 | 630 | |||
XM_008770342.1 | XP_008768564.1 | 4 | 547 | |||
XM_008770343.1 | XP_008768565.1 | 3 | 329 | |||
XM_008770344.1 | XP_008768566.1 | 3 | 470 | |||
GenBank mRNA | BC079253.1 | - | 3 | 378 | 7133170 |
