siRNA Details
Gene Symbol: Hormad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017501.1 | NP_001017501.1 | 2, 3 | 296 | ||
XM_008770340.1 | XP_008768562.1 | 2, 3 | 391 | |||
XM_008770341.1 | XP_008768563.1 | 4, 5 | 548 | |||
XM_008770342.1 | XP_008768564.1 | 3, 4 | 465 | |||
XM_008770343.1 | XP_008768565.1 | 2, 3 | 247 | |||
XM_008770344.1 | XP_008768566.1 | 2, 3 | 388 | |||
GenBank mRNA | BC079253.1 | - | 2, 3 | 296 | 7133170 |
