siRNA Details
Gene Symbol: Zrsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017504.1 | NP_001017504.1 | 1 | 789 | ||
XM_008770435.1 | XP_008768657.1 | 2 | 2708 | |||
XM_008770436.1 | XP_008768658.1 | 2 | 2632 | |||
GenBank mRNA | BC086322.1 | - | 1 | 789 | 7301896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
