siRNA Details
Gene Symbol: Oxsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100508.1 | NP_001093978.1 | 1 | 832 | ||
XM_006251711.1 | XP_006251773.1 | 1 | 615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
