siRNA Details
Gene Symbol: LOC498460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047931.1 | NP_001041396.1 | 3 | 255 | ||
GenBank mRNA | AY383710.1 | - | 3 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
