siRNA Details
Gene Symbol: LOC102547093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251682.2 | XP_006251744.1 | 1 | 190 |
Gene Symbol: LOC102546376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770586.1 | XP_008768808.1 | 1 | 476 | ||
XM_008770587.1 | XP_008768809.1 | 1 | 475 |
Gene Symbol: LOC102546495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770588.1 | XP_008768810.1 | 1 | 89 |
Gene Symbol: LOC102549465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770591.1 | XP_008768813.1 | 1 | 68 | ||
XM_008770592.1 | XP_008768814.1 | 1 | 68 |
Gene Symbol: LOC102550466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770593.1 | XP_008768815.1 | 1 | 381 | ||
XM_008770594.1 | XP_008768816.1 | 1 | 381 |
