siRNA Details
Gene Symbol: Naa30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109099.1 | NP_001102569.1 | 1 | 716 | ||
XM_006251813.2 | XP_006251875.1 | 2 | 1503 |

More Information
Other Species Targeted

Gene Ontology Categories:
