siRNA Details
Gene Symbol: RGD1565222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109101.1 | NP_001102571.1 | 4, 5 | 1379 | ||
XM_006252004.2 | XP_006252066.1 | 4, 5 | 1677 | |||
XM_006252005.2 | XP_006252067.1 | 4, 5 | 1335 | |||
XM_006252006.2 | XP_006252068.1 | 1295 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
