siRNA Details
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170471.1 | NP_001163942.1 | 15 | 4649 | ||
XM_006252316.2 | XP_006252378.1 | 15 | 4705 | |||
XM_006252317.2 | XP_006252379.1 | 15 | 4709 |
