siRNA Details
Gene Symbol: Hesx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109106.1 | NP_001102576.1 | 2 | 669 | ||
XM_008771001.1 | XP_008769223.1 | 4 | 765 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hesx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109106.1 | NP_001102576.1 | 2 | 669 | ||
XM_008771001.1 | XP_008769223.1 | 4 | 765 |