siRNA Details
Gene Symbol: RGD1559804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113787.1 | NP_001107259.1 | 3 | 522 | ||
GenBank mRNA | BC158776.1 | - | 3 | 522 | 7133601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
