siRNA Details
Gene Symbol: RGD1562024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113788.1 | NP_001107260.1 | 2 | 441 | ||
XM_006253581.2 | XP_006253643.1 | 2 | 1323 | |||
XM_006253582.2 | XP_006253644.1 | 2 | 2291 | |||
GenBank mRNA | BC158639.1 | - | 2 | 441 | 7457845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
