siRNA Details
Gene Symbol: LOC102554838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253772.2 | XP_006253834.1 | 5 | 854 |
Gene Symbol: Stmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771561.1 | XP_008769783.1 | 348 |
