siRNA Details
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253849.2 | XP_006253911.1 | 4 | 534 | ||
GenBank mRNA | FQ210031.1 | - | 4 | 406 | ||
FQ227425.1 | - | 4 | 402 | |||
FQ229485.1 | - | 4 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
