siRNA Details
Gene Symbol: RGD1562963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145021.1 | NP_001138493.1 | 4 | 920 | ||
XM_006253780.2 | XP_006253842.1 | 3 | 5213 | |||
GenBank mRNA | BC098763.1 | - | 4 | 920 |
