siRNA Details
Gene Symbol: Tubb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109119.1 | NP_001102589.1 | 673 | |||
XM_008771582.1 | XP_008769804.1 | 698 |

More Information
Other Species Targeted

Gene Ontology Categories:
