siRNA Details
Gene Symbol: RGD1561310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771609.1 | XP_008769831.1 | 1 | 236 | ||
XM_574022.5 | XP_574022.3 | 1 | 236 |
Gene Symbol: LOC100360841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730037.4 | XP_002730083.1 | 1 | 236 |
