siRNA Details
Gene Symbol: LOC102553208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254077.2 | XP_006254139.1 | 4 | 628 | ||
XM_006254078.2 | XP_006254140.1 | 4 | 781 |
Gene Symbol: LOC686142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751717.3 | XP_003751765.1 | 5 | 666 | ||
XM_006254133.2 | XP_006254195.1 | 4 | 1269 | |||
XM_006254135.2 | XP_006254197.1 | 4 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
