siRNA Details
Gene Symbol: Rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031100.1 | NP_112362.1 | 4 | 219 | ||
GenBank mRNA | BC058467.1 | - | 4 | 208 | 6918973 | |
FQ218098.1 | - | 4 | 203 | |||
FQ220555.1 | - | 4 | 202 | |||
FQ222021.1 | - | 4 | 203 | |||
FQ229032.1 | - | 4 | 202 | |||
FQ229696.1 | - | 2 | 330 | |||
FQ233952.1 | - | 4 | 203 | |||
X87106.1 | - | 4 | 219 |
Gene Symbol: RGD1563861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574109.5 | XP_574109.1 | 1 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
