siRNA Details
Gene Symbol: MGC116121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024905.1 | NP_001020076.1 | 3 | 316 | ||
XM_008771953.1 | XP_008770175.1 | 3 | 395 | |||
GenBank mRNA | BC097984.1 | - | 3 | 316 | 7444421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
