siRNA Details
Gene Symbol: Pcdhga12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037337.1 | NP_001032414.1 | 1 | 282 | ||
GenBank mRNA | AY574011.1 | - | 1 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
