siRNA Details
Gene Symbol: RGD1559536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195484.1 | NP_001182413.1 | 4 | 285 | ||
GenBank mRNA | CB779624.1 | - | 4 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
