siRNA Details
Gene Symbol: Fam170a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254732.1 | XP_006254794.1 | 3 | 453 | ||
XM_006254733.2 | XP_006254795.1 | 3 | 447 | |||
XM_006254734.2 | XP_006254796.1 | 3 | 438 | |||
XM_008772162.1 | XP_008770384.1 | 2 | 46 | |||
GenBank mRNA | BC160906.1 | - | 3 | 439 | 7455403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
