siRNA Details
Gene Symbol: RGD1562987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173472.1 | NP_001166943.1 | 2 | 259 | ||
XM_008772163.1 | XP_008770385.1 | 2 | 237 | |||
GenBank mRNA | BC162045.1 | - | 2 | 248 | 7107313 | |
CB568734.1 | - | 2 | 260 | |||
FQ217314.1 | - | 2 | 257 | |||
FQ224078.1 | - | 2 | 247 | |||
FQ228113.1 | - | 2 | 235 | |||
FQ229472.1 | - | 2 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
