siRNA Details
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035257.1 | NP_001030334.1 | 7 | 817 | ||
GenBank mRNA | BC079370.1 | - | 7 | 803 | ||
BC104702.1 | - | 7 | 803 | |||
BC127449.1 | - | 7 | 818 | 8365327 | ||
FQ212517.1 | - | 7 | 822 |
