siRNA Details
Gene Symbol: Arl2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024906.1 | NP_001020077.1 | 3 | 333 | ||
XM_006255120.2 | XP_006255182.1 | 3 | 413 | |||
XM_006255121.2 | XP_006255183.1 | 3 | 441 | |||
GenBank mRNA | BC097448.1 | - | 3 | 333 | 7456585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
