siRNA Details
Gene Symbol: RGD1561415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109129.1 | NP_001102599.1 | 4 | 880 | ||
XM_006255523.2 | XP_006255585.1 | 4 | 876 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1561415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109129.1 | NP_001102599.1 | 4 | 880 | ||
XM_006255523.2 | XP_006255585.1 | 4 | 876 |