siRNA Details
Gene Symbol: Tsnaxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109130.1 | NP_001102600.1 | 4, 5 | 452 | ||
XM_006255527.2 | XP_006255589.1 | 3, 4 | 421 | |||
XM_006255529.2 | XP_006255591.1 | 3, 4 | 282 | |||
XM_008772525.1 | XP_008770747.1 | 2, 3 | 1111 |

More Information
Other Species Targeted

