siRNA Details
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 20 | 4144 | |||
NR_123722.1 | 10 | 2005 | ||||
GenBank mRNA | AK131084.1 | - | 15 | 4508 | ||
AK131085.1 | - | 14 | 4414 | |||
AK302708.1 | - | 10 | 2005 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF132984.1 | - | 2 | 180 | ||
KF175225.1 | - | 2 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
