siRNA Details
Gene Symbol: Fbxo31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044259.2 | NP_001037724.2 | 7 | 882 | ||
XM_006255719.2 | XP_006255781.1 | 7 | 1876 | |||
XM_006255720.2 | XP_006255782.1 | 6 | 1789 | |||
GenBank mRNA | BC110046.1 | - | 5 | 988 | 7302542 | |
BC166840.1 | - | 7 | 882 | 9027777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
