siRNA Details
Gene Symbol: RGD1559896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109134.1 | NP_001102604.1 | 2, 3 | 431 | ||
XM_008772642.1 | XP_008770864.1 | 2, 3 | 448 | |||
XM_008772643.1 | XP_008770865.1 | 2, 3 | 331 | |||
GenBank mRNA | FQ212098.1 | - | 2, 3 | 173 |
