siRNA Details
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 4 | 317 | ||
XM_011522748.2 | XP_011521050.1 | 4 | 414 | |||
XM_011522749.2 | XP_011521051.1 | 5 | 526 | |||
XM_011522750.2 | XP_011521052.1 | 4 | 440 | |||
XM_011522751.1 | XP_011521053.1 | 4 | 413 | |||
XM_017023891.1 | XP_016879380.1 | 5 | 526 | |||
GenBank mRNA | AF132984.1 | - | 4 | 317 | ||
AK160377.1 | - | 23 | 4504 | |||
KF175165.1 | - | 4 | 353 | |||
KF175166.1 | - | 4 | 353 | |||
KF175167.1 | - | 4 | 353 | |||
KF175169.1 | - | 4 | 317 | |||
KF175208.1 | - | 4 | 464 | |||
KF175209.1 | - | 5 | 535 | |||
KF175225.1 | - | 4 | 317 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 4 | 448 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 4 | 317 | ||
XM_005255489.3 | XP_005255546.1 | 6 | 2736 | |||
XM_005255492.4 | XP_005255549.1 | 4 | 721 | |||
XM_006720917.3 | XP_006720980.2 | 6 | 2780 | |||
XM_011522587.1 | XP_011520889.1 | 6 | 2780 | |||
XM_011522593.1 | XP_011520895.1 | 6 | 2780 | |||
XM_011522594.2 | XP_011520896.1 | 6 | 2780 | |||
XM_011522595.1 | XP_011520897.1 | 6 | 2780 | |||
XM_011522596.1 | XP_011520898.1 | 4 | 344 | |||
XM_011522597.1 | XP_011520899.1 | 6 | 2780 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 4 | 317 | ||
XM_005255484.3 | XP_005255541.1 | 5 | 405 | |||
XM_005255485.4 | XP_005255542.1 | 4 | 721 | |||
XM_011522576.2 | XP_011520878.2 | 6 | 2770 | |||
XM_011522577.2 | XP_011520879.2 | 6 | 2691 | |||
XM_011522578.2 | XP_011520880.2 | 4 | 413 | |||
XM_011522580.2 | XP_011520882.2 | 4 | 344 | |||
XM_011522581.1 | XP_011520883.1 | 6 | 2780 | |||
XM_011522583.2 | XP_011520885.1 | 6 | 2780 | |||
XM_011522584.1 | XP_011520886.1 | 6 | 2780 | |||
XM_011522585.1 | XP_011520887.1 | 4 | 344 | |||
XM_011522586.1 | XP_011520888.1 | 6 | 2780 | |||
XM_017023553.1 | XP_016879042.1 | 6 | 2780 | |||
XM_017023554.1 | XP_016879043.1 | 5 | 2693 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 4 | 317 | ||
XM_011522329.2 | XP_011520631.1 | 6 | 728 | |||
XM_011522336.2 | XP_011520638.1 | 6 | 721 | |||
XM_011522337.1 | XP_011520639.1 | 4 | 344 | |||
GenBank mRNA | EL734608.1 | - | 4 | 317 | ||
KF175168.1 | - | 4 | 314 | |||
KF175170.1 | - | 4 | 317 | |||
KF175171.1 | - | 4 | 311 | |||
KF175172.1 | - | 4 | 317 | |||
KF175173.1 | - | 2 | 92 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 4 | 317 | ||
XM_005255735.4 | XP_005255792.1 | 4 | 439 | |||
XM_011522339.2 | XP_011520641.1 | 4 | 365 | |||
XM_017022833.1 | XP_016878322.1 | 5 | 390 | |||
XM_017022834.1 | XP_016878323.1 | 5 | 442 | |||
XM_017022835.1 | XP_016878324.1 | 5 | 503 | |||
XM_017022836.1 | XP_016878325.1 | 4 | 310 | |||
XM_017022837.1 | XP_016878326.1 | 5 | 390 | |||
XM_017022838.1 | XP_016878327.1 | 4 | 785 | |||
XM_017022839.1 | XP_016878328.1 | 5 | 390 | |||
XM_017022840.1 | XP_016878329.1 | 4 | 330 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 4 | 317 | ||
XM_005255738.4 | XP_005255795.1 | 4 | 439 | |||
XM_006720830.3 | XP_006720893.1 | 4 | 1337 | |||
XM_011522342.2 | XP_011520644.1 | 4 | 366 | |||
XM_017022841.1 | XP_016878330.1 | 5 | 390 | |||
XM_017022842.1 | XP_016878331.1 | 5 | 442 | |||
XM_017022843.1 | XP_016878332.1 | 5 | 503 | |||
XM_017022844.1 | XP_016878333.1 | 4 | 310 | |||
XM_017022845.1 | XP_016878334.1 | 5 | 390 | |||
XM_017022846.1 | XP_016878335.1 | 5 | 390 | |||
XM_017022847.1 | XP_016878336.1 | 4 | 330 | |||
GenBank mRNA | BQ101079.1 | - | 4 | 363 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 29 | 5622 | |||
GenBank mRNA | AK124516.1 | - | 24 | 3877 | ||
AK298381.1 | - | 2 | 75 | |||
AK304604.1 | - | 9 | 1322 | |||
BC039707.1 | - | 5 | 636 | 4479933 | ||
BC046145.1 | - | 6 | 728 | |||
KF175174.1 | - | 4 | 353 | |||
KF175210.1 | - | 5 | 596 | |||
KF175211.1 | - | 6 | 673 | |||
KF175212.1 | - | 7 | 486 | |||
KF175214.1 | - | 5 | 535 | |||
KF175219.1 | - | 4 | 503 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 23 | 4410 | |||
NR_123722.1 | 13 | 2271 | ||||
GenBank mRNA | AK131084.1 | - | 18 | 4774 | ||
AK131085.1 | - | 17 | 4680 | |||
AK302708.1 | - | 13 | 2271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
