siRNA Details
Gene Symbol: Cpne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256466.1 | NP_001243395.1 | 6, 7 | 689 | ||
XM_008772319.1 | XP_008770541.1 | 7, 8 | 866 | |||
GenBank mRNA | AM747280.1 | - | 6, 7 | 689 |

More Information
Other Species Targeted

