siRNA Details
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039343.1 | NP_001034432.1 | 8 | 1907 | ||
XM_006227884.2 | XP_006227946.1 | 8 | 1870 | |||
XM_006227885.2 | XP_006227947.1 | 8 | 1875 | |||
XM_006227886.2 | XP_006227948.1 | 8 | 1874 | |||
GenBank mRNA | AY886530.1 | - | 8 | 1907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
