siRNA Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109137.1 | NP_001102607.1 | 7 | 1099 | ||
XM_006227887.2 | XP_006227949.1 | 8 | 3452 | |||
XM_008758736.1 | XP_008756958.1 | 8 | 2936 | |||
XM_008758737.1 | XP_008756959.1 | 8 | 3624 |

More Information
Other Species Targeted

Gene Ontology Categories:
