siRNA Details
Gene Symbol: Sytl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127560.1 | NP_001121032.1 | 2 | 472 | ||
XM_006227888.2 | XP_006227950.1 | 3 | 1025 | |||
XM_006227891.2 | XP_006227953.1 | 3 | 1025 | |||
GenBank mRNA | BC166706.1 | - | 2 | 472 | 5599231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
