siRNA Details
Gene Symbol: Znf761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228009.2 | XP_006228071.1 | 4 | 1238 | ||
XM_006228010.2 | XP_006228072.1 | 5 | 2162 | |||
XM_006228011.2 | XP_006228073.1 | 5 | 2157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
