siRNA Details
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100565.1 | NP_001094035.1 | 7 | 2130 | ||
GenBank mRNA | BC092604.1 | - | 1 | 960 | ||
BC166745.1 | - | 7 | 1963 | 7939059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
