siRNA Details
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170601.1 | NP_001164072.1 | 1804 | |||
XM_008770133.1 | XP_008768355.1 | 2250 | ||||
GenBank mRNA | BC088335.1 | - | 1 | 60 | ||
BC107914.1 | - | 954 | ||||
BC158784.1 | - | 453 | 7302022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
