siRNA Details
Gene Symbol: Zfp94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037212.2 | NP_001032289.2 | 5 | 911 | ||
XM_006228456.2 | XP_006228518.1 | 5 | 1598 | |||
XM_008758964.1 | XP_008757186.1 | 5 | 1527 | |||
XM_008758965.1 | XP_008757187.1 | 5 | 1562 | |||
XM_008758966.1 | XP_008757188.1 | 5 | 1057 | |||
XM_008758967.1 | XP_008757189.1 | 2 | 638 | |||
GenBank mRNA | BC108299.1 | - | 5 | 911 | 7456297 |

More Information
Other Species Targeted

Gene Ontology Categories:
