siRNA Details
Gene Symbol: LOC103690163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759224.1 | XP_008757446.1 | 3 | 1574 | ||
GenBank mRNA | FQ224550.1 | - | 3 | 1502 |
Gene Symbol: Zfp420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728737.4 | XP_002728783.2 | 3 | 1574 |
