siRNA Details
Gene Symbol: LOC102552698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228752.2 | XP_006228814.1 | 2009 |
Gene Symbol: Zfp569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759227.1 | XP_008757449.1 | 1 | 2009 | ||
GenBank mRNA | U78144.1 | - | 1 | 133 |
