siRNA Details
Gene Symbol: Zfp566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134726.1 | NP_001128198.1 | 541 | |||
GenBank mRNA | BC167072.1 | - | 541 | 9028223 |

More Information
Other Species Targeted

Gene Ontology Categories:
