siRNA Details
Gene Symbol: Ndufb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037338.1 | NP_001032415.1 | 3 | 359 | ||
GenBank mRNA | CB732095.1 | - | 3 | 359 | ||
DQ255901.1 | - | 3 | 212 | |||
FQ217067.1 | - | 3 | 366 |

More Information
Other Species Targeted

Gene Ontology Categories:
