siRNA Details
Gene Symbol: MGC114499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024907.1 | NP_001020078.1 | 3 | 889 | ||
GenBank mRNA | BC097443.1 | - | 3 | 889 | 7456163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
