siRNA Details
Gene Symbol: LOC499136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047937.1 | NP_001041402.1 | 2 | 563 | ||
GenBank mRNA | AY383676.1 | - | 2 | 563 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC499136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047937.1 | NP_001041402.1 | 2 | 563 | ||
GenBank mRNA | AY383676.1 | - | 2 | 563 |