siRNA Details
Gene Symbol: LOC102555672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228941.2 | XP_006229003.1 | 4 | 3699 | ||
XM_006228942.2 | XP_006229004.1 | 4 | 7347 | |||
XM_006228943.2 | XP_006229005.1 | 4 | 3707 |
