siRNA Details
Gene Symbol: LOC100911976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228963.2 | XP_006229025.1 | 2 | 273 | ||
XM_008759429.1 | XP_008757651.1 | 2 | 273 |
Gene Symbol: Izumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229184.2 | XP_006229246.1 | 2 | 273 | ||
XM_008759451.1 | XP_008757673.1 | 2 | 273 |
