siRNA Details
Gene Symbol: RGD1562492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109146.1 | NP_001102616.1 | 9 | 1525 | ||
XM_006229119.2 | XP_006229181.1 | 11 | 2288 | |||
GenBank mRNA | BC168918.1 | - | 11 | 1525 | 7131823 |
